Synonym:
2-
CAS Number 6151-25-3
Empirical Formula (Hill Notation) C15H10O7 · 2H2O
Molecular Weight 338.27
Beilstein Registry Number 317313
Colour Index Number 75670
EC Number 204-187-1
MDL number MFCD00149487
PubChem Substance ID 24278668
Popular Documents: Product Information Sheet (PDF) | FTNMR (PDF)
| Related Categories | Acacia, Allium cepa (Onion), Allium sativum (Garlic), Antioxidant and Free Radical Scavengers, Armoracia rusticana (Horseradish), |
| assay | ≥98% (HPLC) |
| form | powder |
| color | yellow |
| mp | >300 °C(lit.) |
| solubility | 1 M NaOH: soluble50 mg/mL |
Live Chat and Frequently Asked Questions
are available for this Product.
Quercetin dihydrate is a flavonoid with anticancer activity. Quercetin is a mitochondrial ATPase and phosphodiesterase inhibitor. It Inhibits PI3-kinase activity and slightly inhibits PIP kinase activity. Quercetin has antiproliferative effects on cancer cell lines, reduces cancer cell growth via type II estrogen receptors, and arrests human leukemic T cells in late G1 phase of the cell cycle.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?