• USA Home
  • Q0134 - Q-Ceramic HyperD® 20

EMAIL THIS PAGE TO A FRIEND
Q0134 Sigma

Q-Ceramic HyperD® 20

20 μm mean particle size

Synonym: HyperD® ceramic ion exchangers

Purchase

Properties

mean particle size   20 μm
capacity   >0.20 meq/mL
storage temp.   2-8°C

Description

Features and Benefits

HyperD media are highly porous, rigid ceramic beads filled with a functionalized hydrophilic gel. The rigid bead allows extremely high linear flow rates without bed compression. The hydrogel exchanges throughout its volume, not just on the surface. And the media do not change volume due to changes in pH or ionic strength.
The supports are stable in commonly used solvents and alkaline or acid environments. Supplied as aqueous suspensions in 1 M NaCl with 20% ethanol to inhibit bacterial growth.

Application

Ceramic HyperD media are used in protein chromatography and affinity chromatography. Ceramic HyperD media have been used in the development of a capture step of a human recombinant F(ab′)(2) produced and expressed in baculovirus-infected cells and to identify whey proteins as a model system for chromatographic separation of proteins.

Legal Information

HyperD is a registered trademark of Pall Corporation

Price and Availability


Amicon Pro Purification System
Safety & Documentation

Safety Information

RIDADR 
UN 1170 3/PG 3
WGK Germany 
3
Flash Point(F) 
105 °F
Flash Point(C) 
40 °C

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?