EMAIL THIS PAGE TO A FRIEND
Q104 Sigma

Quinolinic acid

Synonym: 2,3-Pyridinedicarboxylic acid

  • CAS Number 89-00-9

  • Empirical Formula (Hill Notation) C7H5NO4

  • Molecular Weight 167.12

  •  Beilstein Registry Number 137110

  •  EC Number 201-874-8

  •  MDL number MFCD00006295

  • Popular Documents:  FTNMR (PDF)  

Purchase

Properties

InChI Key   GJAWHXHKYYXBSV-UHFFFAOYSA-N

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
US7967250
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

comparable product

Product #

Description

Add to Cart

P63204 2,3-Pyridinedicarboxylic acid, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?