• USA Home
  • Q107 - N-Methylquipazine dimaleate salt

EMAIL THIS PAGE TO A FRIEND
Q107 Sigma

N-Methylquipazine dimaleate salt

solid

Synonym: 2-[1-(4-Methyl)piperazinyl]quinoline dimaleate salt

  • CAS Number 28614-26-8

  • Empirical Formula (Hill Notation) C14H17N3 · 2C4H4O4

  • Molecular Weight 459.45

  •  MDL number MFCD00153869

  •  PubChem Substance ID 24278115

Purchase

Properties

Related Categories Agonists, Cell Biology, Cell Signaling and Neuroscience, Neuroscience, Neurotransmission,
InChI Key   HNSITEGFVDCKMF-SPIKMXEPSA-N
form   solid
color   white
solubility   0.1 M HCl: soluble
  0.1 M NaOH: slightly soluble
  H2O: soluble
  ethanol: slightly soluble
storage temp.   2-8°C
Gene Information   human ... HTR3A(3359), HTR3B(9177), HTR3C(170572), HTR3D(200909), HTR3E(285242)

Description

Biochem/physiol Actions

Tertiary amine analog of quipazine that is an agonist at 5-HT3 serotonin receptors with an affinity similar to that of quipazine, but which lacks affinity for central 5-HT1B serotonin receptors.

Price and Availability


All labs need water
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?