EMAIL THIS PAGE TO A FRIEND
Q3504 Sigma

Quazinone

powder

Synonym: Ro 13-6438

  • CAS Number 70018-51-8

  • Empirical Formula (Hill Notation) C11H10ClN3O

  • Molecular Weight 235.67

  •  MDL number MFCD00211176

  •  PubChem Substance ID 24278669

Purchase

Description

Biochem/physiol Actions

Phosphodiesterase III (PDE3) inhibitor.

Features and Benefits

This compound is also offered as part of Sigma′s Library of Pharmacologically Active Compounds (LOPAC®1280), a biologically annotated collection of high-quality, ready-to-screen compounds. Click here to learn more.

This compound was developed by Roche. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Legal Information

LOPAC is a registered trademark of Sigma-Aldrich Co. LLC

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?