• USA Home
  • QSC20 - Quantum Simply Cellular Microbead Kit

EMAIL THIS PAGE TO A FRIEND
QSC20 Sigma

Quantum Simply Cellular Microbead Kit

Purchase

Properties

usage   1 kit sufficient for 20 tests
shipped in   wet ice
storage temp.   2-8°C

Description

Specificity

For Quantitative Flow Cytometry. The Quantum Simply Cellular microbeads use flow cytometry to measure the quantity of cell surface antigen molecules in Antibody Binding Capacity (ABC) units. ABC is the number of antibodies bound per cell or microbead. These microbeads are designed for direct or indirect assays which use fluorescent labeled mouse primary antibody or fluorescent-labeled anti-mouse secondary antibody. Four populations of microbeads (approximately the size of lymphocytes) are produced to bind defined, incremental amounts of mouse immunoglobulin. A non-binding microbead population serves as negative control by indicating the detection limit of the cytometer. Microbeads bind staining reagents through covalently-
bound, polyclonal anti-mouse immunoglobulins. This antibody has equivalent reactivity to each mouse isotype (IgG1, IgG2a and IgG2b).
A QuickCal Disk is supplied with every kit, which contains serial numbered folders holding lot specific information (i.e., kit name, assigned calibration values, lot numbers, expiration date). The encrypted information contained in this diskette can be read with the QuickCal v2.1 executable program (sold separately, Stock No. QCAL-10) to generate calibration plots, calculate instrument performance parameters and determine the fluorescence intensity of samples in MESF units of PE.

Legal Information

Quantum Simply Cellular is a trademark of Flow Cytometry Standards Corp.

Price and Availability

Standard component

Description

Product #

Add to Cart

Quantum Simply Cellular Microbeads 1 mL/vial SDS    
QuickCal data diskette 1 ea SDS    
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?