EMAIL THIS PAGE TO A FRIEND
R0126 Sigma

Resin, Khorana

Synonym: Khorana’s Preparation II

Purchase

Properties

Related Categories Biochemicals and Reagents, Nucleosides, Nucleotides, Oligonucleotides, Synthetic Oligonucleotides and Related Reagents More...
extent of labeling   ~100 μmol active chloride per g
storage temp.   −20°C

Description

Application

Used in the synthesis of oligodeoxynucleotides.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?