EMAIL THIS PAGE TO A FRIEND
R0150 Sigma
Piggyback rack
Robust grip, no. of holes, 96, for 0.5 mL tubes
Purchase
Properties
Related Categories Labware, Microcentrifuge Tube Racks, Racks More...
feature   no. of holes 96
Description
General description
The Piggyback Rack is the foam security and storage system that lends a new level of ease to handling microcentrifuge tubes and test tubes, eliminating problems when tubes fall out. Once inserted into a Piggyback Rack, tubes can be easily lifted and stored to free up block racks for long term experimental procedures. The Piggyback Rack will not absorb liquids and is resistant to most solvents, acids and alkalis. They can be marked with a ball point pen or marker on their convenient writing border.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?