EMAIL THIS PAGE TO A FRIEND
R0276 Sigma
NovaRack
purple
Purchase
Properties
Related Categories Centrifuge Tube Racks, Diversified Biotech, Diversified Biotech Racks, Labware, Manufacturer Browser,
mfr. no.   Diversified Biotech
color   purple
Description
General description
NovaRack provides universal housing for test tubes and vials of different sizes on the same surface. The numeric labeling for each tube cavity allows one to easily organize and track up to 46 tubes. It accommodates 24 microcentrifuge tubes (0.5-2 mL), 14 test tubes (12 x 75 mm culture tubes, cryovials and a variety of blood collection tubes), 7 centrifuge tubes (15 mL) and one 50 mL tube. NovaRack is constructed of polypropylene, is autoclavable and is available in five colors. With the addition of the Carousel Extension rod, the space saving ability is doubled and the top rack is allowed to spin.
Legal Information
Novarack is a trademark of Diversified Biotech
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
comparable product
Product #
Description
Add to Cart
R0401 NovaRack, red
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?