• USA Home
  • R0629 - Hpa II from Haemophilus parainfluenzae

EMAIL THIS PAGE TO A FRIEND
R0629 Sigma

Hpa II from Haemophilus parainfluenzae

buffered aqueous glycerol solution

Purchase

Properties

Related Categories 3.1.x.x Acting on esters, 3.x.x.x Hydrolases, Application Index, Biochemicals and Reagents, Enzyme Class Index,
grade   for molecular biology
form   buffered aqueous glycerol solution
concentration   10,000 units/mL
shipped in   wet ice
storage temp.   −20°C

Description

Other Notes

Supplied with 10x Restriction Endonuclease Buffer SL (B3782).

Comment: Hpa II activity is blocked by methylation at either C residue. High salt concentrations (>50 mM potassium chloride) inhibit Hpa II activity.

Linkage

Isoschizomer: Msp I

Specificity

Recognition sequence: 5′-C/CGG-3′
Ligation and recutting results: After 2-10-fold Hpa II overdigestion of 1 μg λ DNA substrate, results in 100% cutting, >95% of fragments can be ligated, and >95% recut.
Heat inactivation: 65 °C for 20 minutes.

Physical form

Solution in 20 mM Tris-HCl, pH 7.5, 0.5 mM EDTA, 50 mM KCl, 5 mM 2-mercaptoethanol, 50% glycerol (v/v), 0.01% polydocanol (v/v), at 4°C

Price and Availability


Biomedical Applications
Safety & Documentation

Safety Information

WGK Germany 
3
RTECS 
RF0757850

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles

Protocols

Restriction Endonucleases - The Molecular Scissors

The term “Restriction enzyme” originated from the studies of Enterobacteria phage λ (lambda phage) in the laboratories of Werner Arber and Matthew Meselson. The ability of certain E. coli strains to ...
Keywords: Asymmetric synthesis, Buffers, Cloning, Degradations, Digestions, Electrophoresis, Gel electrophoresis, Gene expression, Genetic, Indirect Immunofluorescense, Methylations, Molecular biology, Molecular biology techniques, Peptide synthesis, Polymorphisms, Reproduction, Separation, Sequencing

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Related Products

incubation buffer

Product #

Description

Add to Cart

B3782 Restriction Enzyme Buffer SL, 10 ×

related product

Product #

Description

Add to Cart

LIG2 QuickLink DNA Ligation Kit, joins blunt-end and sticky-end DNA fragments
D2886 DNA Ligase from T4-infected Escherichia coli, buffered aqueous glycerol solution

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?