• USA Home
  • R0766 - Microcentrifuge tube rack, Nalgene®
EMAIL THIS PAGE TO A FRIEND
R0766 Sigma
Microcentrifuge tube rack, Nalgene®
capacity, 96 0.5-mL tubes (8 × 12)
Purchase
Properties
Related Categories Labware, Manufacturer Browser, Microcentrifuge Tube Racks, Microcentrifuge tube rack, Nalgene,
material   glass-filled acetal
feature   capacity 96 0.5-mL tubes (8 × 12)
mfr. no.   Nalgene
L × W × H   171 mm × 112 mm × 54 mm
Description
General description
Two-tiered design. Holes in upper and lower tiers offer stable support; wide spacing minimizes interference from overlapping lips. Compatible with most laboratory chemicals, except acids. Autoclavable
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?