EMAIL THIS PAGE TO A FRIEND
R120 Sigma

9-cis-Retinoic acid

Synonym: 9-cis-Tretinoin, Alitretinoin

Purchase

Properties

InChI Key   SHGAZHPCJJPHSC-ZVCIMWCZSA-N
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Articles

UHPLC Analysis of Tretinoin and Related Compounds on Ascentis® Express C18

From our library of Articles, Sigma-Aldrich presents UHPLC Analysis of Tretinoin and Related Compounds on Ascentis® Express C18
Keywords: Chromatography, Pharmaceutical

Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

R4643 9-cis-Retinoic acid, ≥98% (HPLC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?