• USA Home
  • R1253 - Ribonucleic acid, messenger from rabbit globin

EMAIL THIS PAGE TO A FRIEND
R1253 Sigma

Ribonucleic acid, messenger from rabbit globin

lyophilized powder

Synonym: mRNA

Purchase

Properties

Related Categories Biochemicals and Reagents, Nucleosides, Nucleotides, Oligonucleotides, Oligonucleotides, DNA, and RNA More...
form   lyophilized powder
storage temp.   −20°C

Description

Other Notes

Rabbit globin

Physical form

Lyophilized powder containing approx. 35% sodium chloride

Application

Rabbit Globin mRNA is used in a variety of applications including the study of ribosomal structures and protein translation systems. Rabbit Globin mRNA is use to study mechanisms and controls of cDNA biosynthesis.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

92210 Timestrip Plus 0 °C
06693 Timestrip Plus -20 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?