• USA Home
  • R131 - RJR-2429 dihydrochloride

EMAIL THIS PAGE TO A FRIEND
R131 Sigma

RJR-2429 dihydrochloride

solid

Synonym: (±)-2-(3-Pyridinyl)-1-azabicyclo[2.2.2]octane

  • Empirical Formula (Hill Notation) C12H16N2 · 2HCl

  • Molecular Weight 261.19

  •  MDL number MFCD03788206

Purchase

Description

Biochem/physiol Actions

Nicotinic acetylcholine receptor agonist.

Caution

Hygroscopic

Price and Availability


All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?