• USA Home
  • R1392 - Unwire half-racks, Polyketone
EMAIL THIS PAGE TO A FRIEND
R1392 Sigma
Unwire half-racks, Polyketone
capacity, 20 20-mm tubes, blue
Purchase
Properties
Related Categories Labware, Manufacturer Browser, Nalgene, Nalgene Racks, Racks,
material   (ketone resin)
feature   capacity 20 20-mm tubes
mfr. no.   Nalgene
L × W × H   128 mm × 103 mm × 83 mm
color   blue
Description
General description
All the features of the full Unwire racks but more space efficient than full racks in hood, refrigerator, or incubator, when using fewer tubes.
Legal Information
Unwire is a trademark of Thermo Fisher Scientific or its subsidiaries
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?