• USA Home
  • R1506 - Restriction endonuclease from Haemophilus haemolyticus

EMAIL THIS PAGE TO A FRIEND
R1506 Sigma

Restriction endonuclease from Haemophilus haemolyticus

10,000-25,000 units/mL

Purchase

Properties

grade   for molecular biology
form   buffered aqueous solution
concentration   10,000-25,000 units/mL
shipped in   wet ice
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?