• USA Home
  • R1652 - Anti-Rit1 antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
R1652 Sigma

Anti-Rit1 antibody produced in rabbit

~1 mg/mL, fractionated antiserum, buffered aqueous solution

Synonym: Anti-Ras-like protein in tissues

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies for Cell Biology, Antibodies to G Proteins, Antibodies to G Proteins and Cyclic Nucleotides,
species reactivity   human
application(s)   western blot: suitable
clone   polyclonal
concentration   ~1 mg/mL
antibody form   fractionated antiserum
form   buffered aqueous solution
mol wt   antigen mol wt ~36 kDa
shipped in   wet ice
storage temp.   −20°C
Gene Information   human ... RIT1(6016)
biological source   rabbit
conjugate   unconjugated

Description

Immunogen

His-conjugated recombinant human Rit protein.

Antigen Background

Rit, a member of the Ras superfamily of Ras-related GTPases, uses novel effector pathways to regulate proliferation and transformation.

Physical form

Supplied as a 0.2 μm sterile filtered solution in phosphate buffered saline, pH 7.5, containing 0.08% sodium azide.

Price and Availability


Antibody Explorer

Explore with Confidence
Safety & Documentation

Safety Information

WGK Germany  -
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?