• USA Home
  • R1900 - Radioimmunoassay tube racks
EMAIL THIS PAGE TO A FRIEND
R1900 Sigma
Radioimmunoassay tube racks
216 position
Purchase
Properties
Related Categories Antibody/Immuno Supplies and Accessories, Antibody/Immunology Supplies, Labware More...
feature   216 position
Description
Application
These racks provide a practical method of pipetting and storing a large number of samples. By using alternate rows and rotating the tubes, it is easy to keep track of the last tube filled. Also useful in clinical labs for storage of samples.
General description
Steam autoclavable at 121 °C (250 °F).
Price and Availability
Empower your Cancer Research
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?