EMAIL THIS PAGE TO A FRIEND
R2012 Sigma
Reservoirs, glass
capacity 1.4 L
Purchase
Properties
Related Categories Dispensers, Labware, Liquid Handling Equipment, Reagent Reservoirs
mfr. no.   Jencons
availability   not available in EU
capacity   1.4 L
neck thread size   30 mm
Description
General description
These uniquely designed amber reservoirs feature a side entry filler port, allowing the container to be filled without removing the dispenser. Low profile for added stability.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?