• USA Home
  • R2267 - Resin Microscopy And On-Section ImmunoCytoChemistry
EMAIL THIS PAGE TO A FRIEND
R2267 Sigma
Resin Microscopy And On-Section ImmunoCytoChemistry
  •  ISBN-10 0-387-56429-2
  •  ISBN-13 978-0-387-56429-6
Purchase
Properties
Related Categories Books and Software, Immunology Books, Labware More...
publication info   G.R. Newman and J.A. Hobot, Springer-Verlag, 1993, 273 pp., soft cover
mfr. no.   Springer
Description
General description
This manual describes how fixation, resin embedding and immunocytochemistry are interrelated. This book enables non-microscopists to gain a quick grasp of basic principles.
Table of Contents
Part I. Resin Embedding: Strategic Approach.
The Resins.
Resin Embedding Protocols for Chemically Fixed Tissue.
Cryotechniques.
Methods for Resin Polymerisation: Handling Resin Blocks.-
Part II. On-Section Immunolabelling: Strategies in Immunolabelling.
General Considerations.
Immunolabelling Protocols for Resin Sections.
Resin Embedding and Immunolabelling.
List of Suppliers
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?