EMAIL THIS PAGE TO A FRIEND
R2274 Sigma

RANTES from mouse

>95% (SDS-PAGE), recombinant, expressed in E. coli, lyophilized powder, cell culture tested

Purchase

Description

Analysis Note

The biological activity is measured by its ability to chemoattract 2 day cultured human monocytes.

Physical form

Lyophilized from a 0.2 μm filtered solution in 30% acetonitrile and 0.1% trifluoroacetic acid containing 1.25 mg bovine serum albumin.

Price and Availability


All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?