• USA Home
  • R2632 - Reactive Blue 72 Matrix: ::FMCross-linked 4% beaded agarose

EMAIL THIS PAGE TO A FRIEND
R2632 Sigma

Reactive Blue 72 Matrix: ::FMCross-linked 4% beaded agarose

saline suspension

Purchase

Properties

form   saline suspension
extent of labeling   1-3 mg per mL
capacity   1,000-2,000 units/mL binding capacity (malic dehydrogenase)
  3-5 mg/mL binding capacity (human serum albumin)
storage temp.   2-8°C

Description

Physical form

Suspension in 0.5 M NaCl containing 0.02% thimerosal

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?