• USA Home
  • R2901 - Anti-Rab3C antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
R2901 Sigma

Anti-Rab3C antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies for Cell Biology, Antibodies to G Proteins, Antibodies to G Proteins and Cyclic Nucleotides,
species reactivity   rat, mouse, human
application(s)   western blot: suitable
clone   polyclonal
antibody form   affinity isolated antibody
form   buffered aqueous solution
mol wt   antigen mol wt 22 kDa
shipped in   dry ice
storage temp.   −20°C
Gene Information   human ... RAB3C(115827)
mouse ... Rab3c(67295)
rat ... Rab3c(171058)
biological source   rabbit
conjugate   unconjugated

Description

Immunogen

synthetic peptide corresponding to amino acids 9-26 of human RAB3C.

Physical form

Solution in phosphate buffered saline with 1.0 mg/mL bovine serum albumin and 0.05% sodium azide.

Price and Availability


Antibody Explorer

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?