EMAIL THIS PAGE TO A FRIEND
R3012 Sigma

Rac-1 protein human

≥85%, recombinant, expressed in E. coli, buffered aqueous glycerol solution

Purchase

Properties

Related Categories Application Index, Biochemicals and Reagents, Cell Biology, Cell Signaling Enzymes, Cell Signaling and Neuroscience,
recombinant   expressed in E. coli
assay   ≥85%
form   buffered aqueous glycerol solution
mol wt   mol wt 20.5 kDa
shipped in   dry ice
storage temp.   −70°C
Gene Information   human ... RAC1(5879)

Description

Physical form

Solution in 20 mM Tris-HCl, pH 8.0, 45 mM NaCl, 5 mM MgCl2, 1 mM DTT, and 50% glycerol.

Biochem/physiol Actions

GTP-binding protein that is a member of the ras gene superfamily and Rho family of proteins. Rac-1 protein is involved in the regulation of a variety of signaling pathways that control cell growth and differentiation. Rac-1 is suitable as a substrate for geranylgeranyltransferase.

Price and Availability


All labs need water

Biomedical Applications
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
1
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?