• USA Home
  • R3138 - Cryogenic storage rack, stainless steel
EMAIL THIS PAGE TO A FRIEND
R3138 Sigma
Cryogenic storage rack, stainless steel
Vertical; 7-shelf for 51/4 × 51/4 × 33/4 in. boxes
Purchase
Properties
Related Categories Cryogenic Freezer Storage Racks, Cryoware, Labware, Manufacturer Browser, Nalgene,
mfr. no.   Nalgene
overall H   27 3/4 in.
Description
General description
Racks hold CryoBoxes securely but separately, allowing fast retrieval and immediate return to freezer. The vertical racks are best for use in chest freezers, horizontal racks for upright freezers.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?