• USA Home
  • R3143 - Ribonucleic acid, transfer, phenylalanine specific from Escherichia coli MRE 600

EMAIL THIS PAGE TO A FRIEND
R3143 Sigma

Ribonucleic acid, transfer, phenylalanine specific from Escherichia coli MRE 600

lyophilized powder

Synonym: E. coli Phe tRNA

Purchase

Properties

grade   for molecular biology
form   lyophilized powder
storage temp.   −20°C

Description

General description

Transfer RNA is isolated from E. coli MRE 600. This transfer RNA is specific for transfer of the amino acid phenylalanine.

Application

Suitable for use:
• As a tRNA standard
• As a substrate uin tRNA synthatase assays
• As a substrate in other reactions involving tRNAs

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

R4018 Ribonucleic acid, transfer, phenylalanine specific from brewer's yeast
R5636 Ribonucleic acid, transfer from baker's yeast (S. cerevisiae), buffered aqueous solution
R9001 Ribonucleic acid, transfer from baker's yeast (S. cerevisiae), Type X, lyophilized powder
R8508 Ribonucleic acid, transfer from baker's yeast (S. cerevisiae), buffered aqueous solution
R7876 Ribonucleic acid, transfer from wheat germ, Type V, 15-19 unit/mg solid

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?