• USA Home
  • R3266 - Unwire test tube retainers
EMAIL THIS PAGE TO A FRIEND
R3266 Sigma
Unwire test tube retainers
white
Purchase
Properties
mfr. no.   Nalgene
color   white
Description
Compatibility
For 40-position 20 mm test tube rack
General description
For Unwire test tube racks. Snap on top of rack. Molded-in finger tabs hold tubes securely in place on orbital mixers and when inverted.
Legal Information
Unwire is a trademark of Thermo Fisher Scientific or its subsidiaries
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?