• USA Home
  • R3378 - Red blood cells from sheep

EMAIL THIS PAGE TO A FRIEND
R3378 Sigma

Red blood cells from sheep

dry powder (glutaraldehyde treated)

Purchase

Description

Application

Red blood cells from sheep have been used to analyze T-lymphocyte dependent antibody response by ELISA. SRBCs are treated with glutaraldehyde to improve binding of the cells to the assay plate.

Price and Availability


Biomedical Applications
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?