• USA Home
  • R3382 - Anti-Rabbit IgG (whole molecule) antibody produced in goat

EMAIL THIS PAGE TO A FRIEND
R3382 Sigma

Anti-Rabbit IgG (whole molecule) antibody produced in goat

fractionated antiserum, lyophilized powder

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Primary Antibodies, RAB-RAD, Rabbit Secondary Antibodies and Conjugates,
biological source   goat
antibody form   fractionated antiserum
clone   polyclonal
form   lyophilized powder
packaging   vial of 2 mL lyophilized antiserum
application(s)   Ouchterlony double diffusion: 1:16
conjugate   unconjugated
storage temp.   2-8°C

Description

Reconstitution

Reconstitute with 2 ml deionized water.

Physical form

Lyophilized from 0.01 M phosphate buffered saline, pH 7.2

Price and Availability


Antibody Explorer

Explore with Confidence

Amplified Detection
Safety & Documentation

Safety Information

WGK Germany 
3
Protocols & Articles

Articles

How to Choose a Secondary Antibody

The following information is provided to help you decide which secondary antibody may be best for your particular application.
Keywords: Amplification, Enzyme-linked immunosorbent assay, Flow cytometry, Immobilization, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western blot

Secondary Antibodies, Conjugates and Kits

Secondary antibodies are polyclonal or monoclonal antibodies that bind to primary antibodies or antibody fragments, such as the Fc or Fab regions. They are typically labeled with probes that make the...
Keywords: Absorption, Adsorption, Amplification, Bacterial conjugations, Cancer, Digestions, Electrophoresis, Enzyme-linked immunosorbent assay, Flow cytometry, Gel electrophoresis, Gene expression, Hormones, Immobilization, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunology, Immunoprecipitation, Infrared spectroscopy, Microbiology, Microscopy, Purification, Vitamins, Western blot

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Inflammation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Related Products

comparable product

Product #

Description

Add to Cart

R5506 Anti-Rabbit IgG (whole molecule) antibody produced in goat, IgG fraction of antiserum, buffered aqueous solution

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?