• USA Home
  • R3879 - Reactive yellow 2-dextran

EMAIL THIS PAGE TO A FRIEND
R3879 Sigma

Reactive yellow 2-dextran

Purchase

Properties

extent of labeling   100-200 μmol per g
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?