• USA Home
  • R4009 - Rm3 medium need 1250 to add expiration date field

EMAIL THIS PAGE TO A FRIEND
R4009 Sigma

Rm3 medium need 1250 to add expiration date field

Purchase

Properties

Quality Level   GMP
storage temp.   2-8°C

Description

Other Notes

24 month expiration

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?