• USA Home
  • R4253 - BstE II from Bacillus stearothermophilus ET

EMAIL THIS PAGE TO A FRIEND
R4253 Sigma

BstE II from Bacillus stearothermophilus ET

Restriction Enzyme

Purchase

Properties

Related Categories 3.1.x.x Acting on esters, 3.x.x.x Hydrolases, Application Index, Biochemicals and Reagents, Enzyme Class Index,
grade   for molecular biology
form   buffered aqueous glycerol solution
concentration   10,000 units/mL
shipped in   wet ice
storage temp.   −20°C

Description

Other Notes

Supplied with 10x Reaction Buffer BstE II

Comment: BstEII results in 10-15% activity at 37°C.

Specificity

Recognition sequence: 5′-G/GTNACC-3′
Cutting results: a 2-10-fold BstE II overdigestion of 1 μg λ DNA substrate results in 100% cutting
Heat inactivation: This enzyme cannot be inactivated at 65 °C for 15 minutes.

Physical form

Solution in 10 mM Tris-HCl, pH 7.4, 100 mM KCl, 0.1 mM EDTA, 1 mM DTT, 200 μg/ml BSA and 50% [v/v] glycerol

Price and Availability


Biomedical Applications
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
1

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles

Protocols

Restriction Endonucleases - The Molecular Scissors

The term “Restriction enzyme” originated from the studies of Enterobacteria phage λ (lambda phage) in the laboratories of Werner Arber and Matthew Meselson. The ability of certain E. coli strains to ...
Keywords: Asymmetric synthesis, Buffers, Cloning, Degradations, Digestions, Electrophoresis, Gel electrophoresis, Gene expression, Genetic, Indirect Immunofluorescense, Methylations, Molecular biology, Molecular biology techniques, Peptide synthesis, Polymorphisms, Reproduction, Separation, Sequencing

Restriction Enzyme Digest Protocol

Sigma® Life Science’s restriction enzyme collection has been optimized for digestion using five unique buffers. When digesting DNA using a single enzyme, use the buffer supplied with the enzyme (also...
Keywords: Buffers, Digestions, Purification

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Related Products

incubation buffer

Product #

Description

Add to Cart

B8781 Restriction Enzyme Buffer SB, 10 ×

related product

Product #

Description

Add to Cart

LIG2 QuickLink DNA Ligation Kit, joins blunt-end and sticky-end DNA fragments
R0260 BamH I from Bacillus amyloliquefaciens H, Restriction Enzyme
R7260 Xba I from Xanthomonas badrii, Restriction Enzyme
D2886 DNA Ligase from T4-infected Escherichia coli, buffered aqueous glycerol solution
R1137 Hind III from Haemophilus influenzae, Restriction Enzyme
R2756 EcoR V from Escherichia coli, buffered aqueous glycerol solution

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?