• USA Home
  • R4261 - Centrifuge tube rack, acrylic
EMAIL THIS PAGE TO A FRIEND
R4261 Sigma
Centrifuge tube rack, acrylic
15-position, for 15 mL plastic centrifuge tubes
Purchase
Properties
Related Categories Centrifuge Tube Racks, Labware, Racks More...
Description
General description
Acrylic, S-profile with non-skid feet. Overall dimensions: 16.3 × 7.5 × 9 cm. Three racks fit comfortably inside the large acrylic β-storage container, product No. S4144.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
comparable product
Product #
Description
Add to Cart
Z675970 Rotating 4-way racks, Large tubes
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?