• USA Home
  • R4266 - Unwire full racks, polypropylene
EMAIL THIS PAGE TO A FRIEND
R4266 Sigma
Unwire full racks, polypropylene
capacity, 72 16-mm tubes (6 × 12), green
Purchase
Properties
Related Categories Labware, Manufacturer Browser, Nalgene, Nalgene Racks, Racks,
material   polypropylene
feature   capacity 72 16-mm tubes (6 × 12)
mfr. no.   Nalgene
L × W × H   248 mm × 127 mm × 70 mm
color   green
Description
General description
Unwire test tube racks have no coating that can chip or peel and permit corrosion. Excellent replacement for wire racks; withstands most lab chemicals. Excellent thermal properties and won′t get brittle in freezers. Grid design allows tubes to be tipped from side to side for clearly marking position. These racks will not float in water baths; do not autoclave polypropylene racks.
Legal Information
Unwire is a trademark of Thermo Fisher Scientific or its subsidiaries
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?