• USA Home
  • R4636 - Microcentrifuge tube rack
EMAIL THIS PAGE TO A FRIEND
R4636 Sigma
Microcentrifuge tube rack
insert holds, 72 0.5-mL tubes
Purchase
Properties
Related Categories Labware, Microcentrifuge Tube Racks, Racks More...
material   expanded polystyrene
feature   insert holds 72 0.5-mL tubes
packaging   case of 30 complete racks
L × W × H   7 1/4 in. × 4 5/16 in. × 2 5/8 in.
Description
General description
Unique three-part design (box, drilled insert, and cover) provides microcentrifuge tube storage and more.
• Insert can be floated in a water bath for incubation of tubes
• Insulating box can be filled with liquid nitrogen, dry or wet ice for cooling or flash freezing
• Useful for sample transportation because of compactness, insulating properties, and lightweight
Manufacturing process is environmentally safe.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?