• USA Home
  • R4751 - Anti-Rabbit Serum antibody produced in goat

EMAIL THIS PAGE TO A FRIEND
R4751 Sigma

Anti-Rabbit Serum antibody produced in goat

fractionated antiserum, lyophilized powder

Synonym: Anti-Serum, rabbit

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies to Animal Plasma Proteins, Primary Antibodies, RAB-RAD,
biological source   goat
antibody form   fractionated antiserum
clone   polyclonal
form   lyophilized powder
species reactivity   rabbit
packaging   vial of 2 mL lyophilized antiserum
application(s)   immunoelectrophoresis: suitable
conjugate   unconjugated
storage temp.   2-8°C

Description

Reconstitution

Reconstitute with 2 mL deionized water.

Physical form

Lyophilized from 0.01 M phosphate buffered saline, pH 7.2

Price and Availability


Antibody Explorer

Explore with Confidence
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Inflammation, Molecular biology, Phosphorylations, Purification, Western blot

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?