• USA Home
  • R4768 - Reticulum TISSUE-TROL

EMAIL THIS PAGE TO A FRIEND
R4768 Sigma

Reticulum TISSUE-TROL

from human liver tissue containing reticular fibers

Purchase

Properties

Related Categories Cell Biology, Hematology and Histology, TISSUE-TROL Control Slides More...
Quality Level   GMP-IVD
biological source   from human liver tissue containing reticular fibers
shelf life   Expiry date on the label.
IVD   for in vitro diagnostic use

Description

Components

25 slides per box. 1 stained slide is provided with each box. The stained slide has been stained with Sigma Procedure No. HT102.

Application

Suitable for use with Sigma Reticulum Stain Procedure No. HT102 and other silver impregnation techniques.

Legal Information

TISSUE-TROL is a trademark of Sigma-Aldrich Co. LLC

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

TTR010 Reticulum TISSUE-TROL Control Slides

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?