• USA Home
  • R5275 - Anti-RAIDD antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
R5275 Sigma

Anti-RAIDD antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym: Anti-CRADD

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies for Apoptosis, Antibodies for Cell Biology, Antibodies to Adaptors,
species reactivity   human
application(s)   microarray: suitable
  western blot: 1 μg/mL using human HeLa cell extract
clone   polyclonal
antibody form   IgG fraction of antiserum
form   buffered aqueous solution
mol wt   antigen mol wt 22 kDa
shipped in   dry ice
storage temp.   −20°C
Gene Information   human ... CRADD(8738)
biological source   rabbit
conjugate   unconjugated

Description

Immunogen

synthetic peptide corresponding to amino acids 181-199 of the C-terminal end of human RAIDD.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide.

Specificity

The antibody reacts with RAIDD (RIP-Associated ICH-1/CED-3-homologous protein with DD). This protein is also termed CRADD (Caspase and RIP Adaptor with DD).

Price and Availability


Antibody Explorer

Explore with Confidence
Safety & Documentation

Safety Information

WGK Germany  3
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?