• USA Home
  • R5400 - Anti-RICK, N-Terminal antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
R5400 Sigma

Anti-RICK, N-Terminal antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym: Anti-CARDIAK, Anti-RIP2

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies for Apoptosis, Antibodies for Cell Biology, Antibodies for Kinase/Phosphatase Biology,
species reactivity   human
application(s)   ELISA: suitable
  immunocytochemistry: suitable
  immunofluorescence: suitable
  western blot: suitable
clone   polyclonal
antibody form   affinity isolated antibody
form   buffered aqueous solution
mol wt   antigen mol wt 59 kDa
shipped in   dry ice
storage temp.   −20°C
Gene Information   human ... RIPK2(8767)
biological source   rabbit
conjugate   unconjugated

Description

Immunogen

synthetic peptide corresponding to amino acids 11-30 of the N-terminal end of human RICK (RIP-like Interacting CLARP Kinase)

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide.

Price and Availability


Antibody Explorer

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?