• USA Home
  • R5907 - Anti-RUVBL1 (161-175) antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
R5907 Sigma

Anti-RUVBL1 (161-175) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym: Anti-ECP54, Anti-INO80H, Anti-NMP238, Anti-PONTIN, Anti-Pontin52, Anti-RVB1, Anti-RuvB-like 1 isoform 1, Anti-TIH1, Anti-TIP49, Anti-TIP49A

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies to Transcription Factors, Epigenetics, Primary Antibodies,
species reactivity   human
application(s)   western blot: 1:500-1:2,000
clone   polyclonal
antibody form   IgG fraction of antiserum
form   buffered aqueous solution
mol wt   antigen mol wt ~50 kDa
shipped in   dry ice
storage temp.   −20°C
Gene Information   human ... RUVBL1(8607)
biological source   rabbit
conjugate   unconjugated

Description

Immunogen

synthetic peptide corresponding to amino acids 161-175 of human RUVBL1

General description

RUVBL1 (RuvB-like AAA ATPase 1) is a 50kDa protein belonging to the AAA(+)-family of ATPases (ATPase associated with diverse cellular activities). It is composed of three functional domains, named as (DI, DII, and DIII domains. DII domain consists of 6 α-helices, 9 β-strands, and 2 very short 310-helices. Among all the domains, two domains are responsible for ATP binding and hydrolysis.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Biochem/physiol Actions

RUVBL1 (RuvB-like AAA ATPase 1) is involved in various cellular processes including transcriptional regulation, DNA replication, DNA damage repair, chromatin remodeling and apoptosis. In the chromatin-remodeling process, it conjugates with other proteins to form the human histone acetylase/chromatin-remodeling complex TIP60, which plays an essential role in transcription and DNA repair. The chromatin-remodeling complex regulates chromatin structure as well as it maintains DNA-based export in the cell. It also functions as a component of the human RNA polymerase II holoenzyme complex, which indicates its role in transcriptional processes. RUVBL1 is also associated with two oncogenic pathways, c-Myc and another, β-catenin pathways. In addition, it is also involved in small nucleolar ribonucleoprotein particle assembly, nucleolar localization, and trafficking.

Price and Availability


Antibody Explorer

Explore with Confidence
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?