• USA Home
  • R6143 - Reactive Red 120−Agarose

EMAIL THIS PAGE TO A FRIEND
R6143 Sigma

Reactive Red 120−Agarose

saline suspension

Purchase

Properties

Related Categories Affinity Chromatography, Dye Resins, Molecular Biology, Protein Chromatography, Proteomics More...
form   saline suspension
extent of labeling   7-9 μmol per mL
matrix   fast flow highly cross-linked 6% agarose
matrix attachment   triazine
capacity   ≥4,000 units/mL binding capacity (lactic dehydrogenase)
storage temp.   2-8°C

Description

Physical form

Suspension in 0.5 M NaCl containing 0.02% thimerosal

Price and Availability


Amplified Detection
Safety & Documentation

Safety Information

WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?