• USA Home
  • R6383 - Packing reservoirs for jacketed LC columns
EMAIL THIS PAGE TO A FRIEND
R6383 Sigma
Packing reservoirs for jacketed LC columns
size 150 mL, column I.D. 1.0 cm
Purchase
Description
General description
Supplied complete with all fittings necessary for connection to threaded columns. The top has a 1/4 -28 fitting.
Price and Availability
All labs need water
Non-Sterile Millex Syringe Filters
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?