• USA Home
  • R6519 - Ribonucleic acid, transfer, glutamic acid specific II from Escherichia coli

EMAIL THIS PAGE TO A FRIEND
R6519 Sigma

Ribonucleic acid, transfer, glutamic acid specific II from Escherichia coli

lyophilized powder

Purchase

Properties

Related Categories Biochemicals and Reagents, Nucleosides, Nucleotides, Oligonucleotides, Oligonucleotides, DNA, and RNA More...
form   lyophilized powder
storage temp.   −20°C

Description

Quality

Acceptor Activity: ≥1200 picomoles glutamic acid per A260 unit.

Application

Ribonucleic acid, transfer, glutamate-specific may be used as a substrate to study the specifity and kinetics of natural, mutated and chemically engineered glutamate-specific E. coli aminoacyl-tRNA synthetase(s).

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?