• USA Home
  • R7018 - Eppendorf® Microcentrifuge accessories
EMAIL THIS PAGE TO A FRIEND
R7018 Sigma
Eppendorf® Microcentrifuge accessories
Fixed Angle Rotor with lid, (Rotor F-45-24-11), for use with 5415 Microcentrifuge
Purchase
Properties
Related Categories Centrifugation Supplies, Centrifuges, Eppendorf, Eppendorf Centrifugation, Eppendorf Centrifuge parts & Accessories,
material   aluminum (45 degree angle bore)
  polypropylene lid
mfr. no.   Eppendorf®, 5425 725.000 (European)
Description
General description
24-position standard rotor for 1.5/2.0 mL tubes (adapters available for 0.2 mL, 0.4 mL, 0.5 mL and 0.6 mL tubes); autoclavable
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Price and Availability
Steriflip Filters Units
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?