• USA Home
  • R7250 - Ribonucleic acid from calf liver

EMAIL THIS PAGE TO A FRIEND
R7250 Sigma

Ribonucleic acid from calf liver

Type IV

Purchase

Properties

Related Categories Biochemicals and Reagents, Nucleosides, Nucleotides, Oligonucleotides, Oligonucleotides, DNA, and RNA More...
type   Type IV
storage temp.   −20°C

Description

Packaging

100, 250 mg in poly bottle

500 mg in glass bottle

Preparation Note

Prepared by procedure of Kirby, K.S., Biochem. J., 64, 405 (1956).

Application

Ribonucleic acid (RNA) from calf liver may be used as a substrate for studying ribonuclease activities of enzymes such as RNAase(s). RNA from calf liver may also be used as a substrate to study the effects of various reactive nitrogen species on nucleotides.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

92210 Timestrip Plus 0 °C
06693 Timestrip Plus -20 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?