EMAIL THIS PAGE TO A FRIEND
R7401 Sigma
PCR 96 Well rack
yellow
Purchase
Properties
Related Categories Labware, Molecular Biology Supplies, PCR (Polymerase Chain Reaction) Supplies, PCR Tube Racks
material   polypropylene
color   yellow
Description
General description
Sturdy rack for working with 0.2 mL tubes. Accommodates the standard 8 or 12 tube strips. Perfect for pre- or post-PCR sampling applications. The tube wells are easily identified with imprinted numbers and letters. The rack features a removable hinged lid.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?