EMAIL THIS PAGE TO A FRIEND
R8008 Sigma

Fluorescent rulers

visible at 300 and 350 nm

Purchase

Description

General description

Two different intensities match different fluorescent samples. Clearly visible at 300 and 350 nm.

Packaging

Pkg. contains two 7 × 200 mm and two 7 × 100 mm rulers.

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?