• USA Home
  • R8149 - Rhizoxin from Rhizopus sp.

EMAIL THIS PAGE TO A FRIEND
R8149 Sigma

Rhizoxin from Rhizopus sp.

>95% (HPLC)

Purchase

Description

Biochem/physiol Actions

An antitumor agent, rhizoxin is a 16-member ring lactone having an oxazole ring in its structure. This macrolide inhibits microtubule assembly and also depolymerizes pre-formed microtubles.
Rhizoxin binds to β-tubulin in most eucaryotic cells including animals, plants and fungi. It completely prevents formation of an intrachain cross-link in β-tubulin by N,N′-ethylene­bis­(iodo­acetamide). Due to its antimitotic activity it is used as an antitumor agent e.g., in human small cell lung cancer cell lines.

Price and Availability


All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
VI1800000
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?