EMAIL THIS PAGE TO A FRIEND
R8500 Sigma

Methyl ricinelaidate

≥99% (GC), liquid

Synonym: Methyl trans-12-hydroxy-9-octadecenoate, Ricinelaidic acid methyl ester

  • CAS Number 7706-01-6

  • Empirical Formula (Hill Notation) C19H36O3

  • Molecular Weight 312.49

  •  Beilstein Registry Number 1728452

  •  MDL number MFCD00069621

  •  PubChem Substance ID 24899406

Purchase

Properties

Related Categories Biochemicals and Reagents, Esters, Fatty Acids and conjugates, Fatty Acyls, Lipids,
InChI Key   XKGDWZQXVZSXAO-LVMGUKCRSA-N
assay   ≥99% (GC)
form   liquid
storage temp.   −20°C

Description

Packaging

Sealed ampule

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?