• USA Home
  • R8628 - Riboflavin Binding Protein from chicken egg white

EMAIL THIS PAGE TO A FRIEND
R8628 Sigma

Riboflavin Binding Protein from chicken egg white

apo-form, lyophilized powder

Synonym: RBP

Purchase

Properties

Related Categories Auxiliary Proteins, Biochemicals and Reagents, Other, Proteins and Derivatives
form   lyophilized powder
usage   1 mg sufficient for 20 riboflavin determinations
impurities   riboflavin and salts, essentially free
capacity   5-15 μg/mL, protein binding capacity (Riboflavin)
storage temp.   −20°C

Description

Application

Useful in the fluorimetric determination of riboflavin

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?